Learn More
AdipoGen Alternariol monomethyl ether
SDP
Supplier:  AdipoGen BVT0323C500
Encompass_Preferred
Chemical. CAS 26894-49-5. Formula C15H12O5. MW 272.3. Isolated from Alternaria sp. Mycotoxin. Antifungal. Phytotoxic. Shown to possess DNA strand-breaking activity. Inhibits the activity of topoisomerase IIalpha Topo IIalpha in mammalian cells. Antiviral Herpes simplex. Hepatitis C NS3-4A protease inhibitor.
Catalog No. 50-256-1989