Learn More
Abcam Alternariol monomethyl ether, antifungal phytoto x ic compound, 1MG
Catalog No. 502706012
Encompass_Preferred
MW 272.25 Da, Purity >98%. Antifungal phytotoxic compound. Inhibits electron transport chain in chloroplasts. Possesses DNA strand-breaking activity. Inhibits topoisomerase IIα activity. Active in vitro. See similar compound .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6012 Supplier Abcam Supplier No. AB1441761MG
MW 272.25 Da, Purity >98%. Antifungal phytotoxic compound. Inhibits electron transport chain in chloroplasts. Possesses DNA strand-breaking activity. Inhibits topoisomerase IIα activity. Active in vitro. See similar compound .
The product is subject to the following: Abcam Restricted Use Statement