Learn More
CPC Scientific 5FAM-Pro-Leu-Ser-Arg-Thr-Leu-Ser-Val-Ser-Ser-Leu-Pro-Gly-Leu-NH2 (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific PROK006A
Encompass
FAM-labeled Alternate Sytide 2 peptide (Ab/Em = 494/521) that is related to the NH2 terminal of glycogen synthase, a substrate for CAMK and is phosphorylated by DCLK (doublecortin like kinase). ONE-LETTER SEQUENCE: 5FAM-PLSRTLSVSSLPGL-NH2MOLECULAR FORMULA: C84H122N18O25MOLECULAR WEIGHT:1784STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES: Bart, M. et al. Mol. Brain Res. 120, 103 (2004)
Catalog No. 50-211-0227