Learn More
Bachem Fmoc-Tyr(PO(OBzl)OH)-OH
SDP
Catalog No. 501947355
Encompass_Preferred
Fmoc-Tyr(PO(OBzl)OH)-OH, 191348-16-0, C31H28NO8P, Mr 573.54, 5g. Alternative to Fmoc-Tyr(PO3(MDPSE)2)-OH.
Catalog No. 50-194-7355 Supplier Bachem Supplier No. 4038299.0005
Fmoc-Tyr(PO(OBzl)OH)-OH, 191348-16-0, C31H28NO8P, Mr 573.54, 5g. Alternative to Fmoc-Tyr(PO3(MDPSE)2)-OH.