Learn More
Cayman Chemical AltertoxIn I 1mg
SDP
Catalog No. 502983722
Encompass_Preferred
A natural mycotoxin that shows mutagenic activity against S. typhimurium and inhibits human topoisomerase II and bacterial gyrase; completely inhibits replication of HIV-1 virus at 2.2 µM in a cell-based assay
Catalog No. 50-298-3722 Supplier Cayman Chemical Supplier No. 211291 MG
A natural mycotoxin that shows mutagenic activity against S. typhimurium and inhibits human topoisomerase II and bacterial gyrase; completely inhibits replication of HIV-1 virus at 2.2 µM in a cell-based assay