Learn More
TARGETMOL CHEMICALS INC ALTIRATINIB 25MG
SDP
Catalog No. 502577239
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Altiratinib (DCC-2701)(DCC-2701) is a novel c-MET/TIE-2/VEGFR inhibitor; effectively reduce tumor burden in vivo and block c-MET pTyr(1349)-mediated signaling cell growth and migration as compared with an HGF antagonist in vitro. purity: 99%
Catalog No. 50-257-7239 Supplier TARGETMOL CHEMICALS INC Supplier No. T205425MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Altiratinib (DCC-2701)(DCC-2701) is a novel c-MET/TIE-2/VEGFR inhibitor; effectively reduce tumor burden in vivo and block c-MET pTyr(1349)-mediated signaling cell growth and migration as compared with an HGF antagonist in vitro. purity: 99%