Learn More
CERILLO INC Alto Plate Reader, 600nm
Catalog No. NC2603345
Encompass_Preferred
The Alto, 600nm includes one Alto plate reader, one USB cable, one power adapter, one 16 GB microSD card, and 1 year of firmware upgrades.
Catalog No. NC2603345 Supplier CERILLO INC Supplier No. ALT1600
Add to Cart
Add to Cart
Request a Quote
The Alto, 600nm includes one Alto plate reader, one USB cable, one power adapter, one 16 GB microSD card, and 1 year of firmware upgrades.