Learn More
CERILLO INC Alto Shaking Adapter
Catalog No. NC2603346
Encompass_Preferred
The Cerillo Alto Shaking Adapter allows the Alto to be secured to a universal shaker plate via screws and also features high-friction silicone strips and powerful magnets. The adapter features a plate-size extrusion allowing it to fit securely to small shakers designed for a microplate.
Catalog No. NC2603346 Supplier CERILLO INC Supplier No. ALTAPT
The Cerillo Alto Shaking Adapter allows the Alto to be secured to a universal shaker plate via screws and also features high-friction silicone strips and powerful magnets. The adapter features a plate-size extrusion allowing it to fit securely to small shakers designed for a microplate.