Learn More
Medchemexpress LLC HY-B0521 100mg Medchemexpress, Altrenogest CAS:850-52-2 Purity:>98%
SDP
Catalog No. 502082578
Encompass_Preferred
Medchemexpress, HY-B0521 100mg Altrenogest CAS:850-52-2 Altrenogest (Allyltrenbolone) is a progestogen structurally related to veterinary steroid trenbolone. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2578 Supplier Medchemexpress LLC Supplier No. HYB0521100MG
Medchemexpress, HY-B0521 100mg Altrenogest CAS:850-52-2 Altrenogest (Allyltrenbolone) is a progestogen structurally related to veterinary steroid trenbolone. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.