Learn More
TARGETMOL CHEMICALS INC Altrenogest 200MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T0870200MG
Encompass_Preferred
Also available in 1 mL, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Altrenogest (Allyltrenbolone)(A35957; RU2267), a progestogen, has structural relation with veterinary steroid trenbolone. Purity 99.61%
Catalog No. 50-288-0653