Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Altrenogest 98.0+%, TCI America™
SDP

Catalog No. A2892200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2892200MG 200 mg
A28921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A2892200MG Supplier TCI America Supplier No. A2892200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 850-52-2
Molecular Formula C21H26O2
Molecular Weight (g/mol) 310.437
MDL Number MFCD00867855
InChI Key VWAUPFMBXBWEQY-MXEMCNAFSA-N
Synonym 17-alpha-Allyl-17-beta-hydroxyestra-4,9,11-trien-3-one, Allyltrenbolone
PubChem CID 129317581
IUPAC Name (8S,13S,14R,17S)-17-hydroxy-13-methyl-17-prop-2-enyl-1,2,6,7,8,14,15,16-octahydrocyclopenta[a]phenanthren-3-one
SMILES CC12C=CC3=C4CCC(=O)C=C4CCC3C1CCC2(CC=C)O

Specifications

Melting Point 118°C
Color Yellow
Quantity 200 mg
Formula Weight 310.44
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Altrenogest
RTECSNumber KG7745000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.