Learn More
Apexbio Technology LLC Altretamine 645-05-6 10mM (in 1mL Ethanol)
SDP
Catalog No. 5000063553
Encompass_Preferred
Altretamine (CAS 645-05-6) is a small molecule compound characterized primarily by its antineoplastic properties As an antitumor agent it is thought to exert cytotoxic effects through the formation of DNA-alkylating reactive intermediates disrupting DNA synthesis and function thereby inhibiting tumor cell proliferation Beyond oncological research altretamine demonstrates chemosterilant activity leading to sterility in male insects such as houseflies suggesting potential utility in entomological studies exploring population control Altretamine serves as a valuable tool in biomedical research focused on oncology and chemotherapy mechanism exploration
Catalog No. 50-000-63553 Supplier Apexbio Technology LLC Supplier No. A25595.1
Altretamine (CAS 645-05-6) is a small molecule compound characterized primarily by its antineoplastic properties As an antitumor agent it is thought to exert cytotoxic effects through the formation of DNA-alkylating reactive intermediates disrupting DNA synthesis and function thereby inhibiting tumor cell proliferation Beyond oncological research altretamine demonstrates chemosterilant activity leading to sterility in male insects such as houseflies suggesting potential utility in entomological studies exploring population control Altretamine serves as a valuable tool in biomedical research focused on oncology and chemotherapy mechanism exploration