Learn More
Apexbio Technology LLC Altretamine, 500mg Cas# 645-05-6
SDP
Supplier:  Apexbio Technology LLC A2559500
Encompass_Preferred
Altretamine is an antitumor agent which also acts as a chemosterilant for male houseflies and other insects. Altretamine is also an antineoplastic. Other sizes are available. Please inqury us for quote.
Catalog No. 50-199-8514