Learn More
Cayman Chemical Altretamine, Purity: ≥98%, CAS: 645-05-6, 50 mg
SDP
Catalog No. 502995023
Encompass_Preferred
An anticancer agent; induces cytotoxicity in an ovarian cancer cell line at 10 μg/ml; inhibits GPX4 and induces accumulation of lipid-reactive oxygen species in U-2932 cells without depleting GSH levels at 0.5 mM; reduces tumor growth in an M5076 murine sarcoma model at 150 mg/kg
Catalog No. 50-299-5023 Supplier Cayman Chemical Supplier No. 2366250 MG
An anticancer agent; induces cytotoxicity in an ovarian cancer cell line at 10 μg/ml; inhibits GPX4 and induces accumulation of lipid-reactive oxygen species in U-2932 cells without depleting GSH levels at 0.5 mM; reduces tumor growth in an M5076 murine sarcoma model at 150 mg/kg