Learn More
Sorbent Technologies Inc Alumina, Basic, Act. I, 50-200um, 500g
SDP
Catalog No. 501138765
Encompass_Preferred
Alumina Basic, Standard Activity I is a good all-purpose adsorbent for laboratory as well as process use.It has a pH of 10, a particle distribution of 50-200um, and a quantity of 500g. The starting activity level of Alumina, Standard Activity I is about half as high as Alumina, Activity Super I and can be deactivated by the addition of polar media (water, alcohols, etc.) according to the deactivation scale. Standardized manufacturing processes provide a specific Brockmann dryness level for lot to lot reproducibility and precise deactivation possibilities. The Loss on Ignition (LOI) is 1.7 +/- 0.2% and the specific surface is ~150 m2/g. The adsorption behavior is extremely constant. Alumina Basic, Standard Activity I may be used for the purification of solvents. Applications include: Alkaloids, essential oils, plant extraction, purification of organic solvents, insecticides, clarification of fatty oils, removal of alcohol from chloroform
Catalog No. 50-113-8765 Supplier Sorbent Technologies Inc Supplier No. 1564005
Alumina Basic, Standard Activity I is a good all-purpose adsorbent for laboratory as well as process use.It has a pH of 10, a particle distribution of 50-200um, and a quantity of 500g. The starting activity level of Alumina, Standard Activity I is about half as high as Alumina, Activity Super I and can be deactivated by the addition of polar media (water, alcohols, etc.) according to the deactivation scale. Standardized manufacturing processes provide a specific Brockmann dryness level for lot to lot reproducibility and precise deactivation possibilities. The Loss on Ignition (LOI) is 1.7 +/- 0.2% and the specific surface is ~150 m2/g. The adsorption behavior is extremely constant. Alumina Basic, Standard Activity I may be used for the purification of solvents. Applications include: Alkaloids, essential oils, plant extraction, purification of organic solvents, insecticides, clarification of fatty oils, removal of alcohol from chloroform