Learn More
Sorbent Technologies Inc Alumina, Basic (pH 10), Act. II-III, 50-200um, 500g
SDP
Catalog No. 501138744
Encompass_Preferred
Alumina Basic Activity II-III has a pH of about 10, a particle distribution of 50-200um, and quantity of 500g. This alumina is a medium active adsorbent for use with easily adsorbable substances and for the cleavage of alkaloid salts.
Catalog No. 50-113-8744 Supplier Sorbent Technologies Inc Supplier No. 1530005
Alumina Basic Activity II-III has a pH of about 10, a particle distribution of 50-200um, and quantity of 500g. This alumina is a medium active adsorbent for use with easily adsorbable substances and for the cleavage of alkaloid salts.