Learn More
Sorbent Technologies Inc Alumina, Basic, Super I, 50-200um, 500g
SDP
Catalog No. 501138740
Encompass_Preferred
Alumina Basic Super I has a pH of 10, a particle distribution of 50-200um, and quantity of 500g. In a non-polar environment Alumina Super I shows adsorption capacity approximately twice as high as Alumina Activity I. Alumina Basic Super I has a Loss on Ignition (LOI) of 1% and a specific surface area of about 200 m2/g. The high activity level lends itself to the separation of polar samples in nonpolar solvent systems, specifically, the purification of solvents. Applications include: Alkaloids, Synthetic bases, dehydration of ethers, Removal of peroxides, Natural and synthetic dyestuffs, Hydrocarbons
Catalog No. 50-113-8740 Supplier Sorbent Technologies Inc Supplier No. 1516005
Alumina Basic Super I has a pH of 10, a particle distribution of 50-200um, and quantity of 500g. In a non-polar environment Alumina Super I shows adsorption capacity approximately twice as high as Alumina Activity I. Alumina Basic Super I has a Loss on Ignition (LOI) of 1% and a specific surface area of about 200 m2/g. The high activity level lends itself to the separation of polar samples in nonpolar solvent systems, specifically, the purification of solvents. Applications include: Alkaloids, Synthetic bases, dehydration of ethers, Removal of peroxides, Natural and synthetic dyestuffs, Hydrocarbons