Learn More
Crescent Chemical Co Inc ALU-GEL-S SUSPENSION STERILE
SDP
Supplier:  Crescent Chemical Co Inc SE12261.01
Encompass_Preferred
Alu-Gel-S suspension research grade sterile Ph. Eur. 1.3 % in water. Pyrogen free (as assayed in the supernatant), aged, salt-free. Size - 50ML Storage Conditions - +15 °C TO +30 °C Catalog Number - 12261.01
Catalog No. 50-247-277