Learn More
New England Biolabs, Inc. AluI Methyltransferase – 100 units
SDP
Catalog No. 50811640
Encompass
AluI Methyltransferase modifies the cytosine residue (C5) of the sequence AGCT.
Catalog No. 50-811-640 Supplier New England Biolabs, Inc. Supplier No. M0220S
AluI Methyltransferase modifies the cytosine residue (C5) of the sequence AGCT.