Learn More
Sorbent Technologies Inc Alumina, Neutral, Active, 32-63um, 500g
SDP
Catalog No. 501138750
Encompass_Preferred
Alumina Neutral, Active, is used for packing flash chromatography columns. It has a particle distribution of 32-63um and a quantity of 500g. The approximate pH is 7.4, Loss on Ignition (LOI) is < 1.7%, and specific surface area is about 200 m2/g.
Catalog No. 50-113-8750 Supplier Sorbent Technologies Inc Supplier No. 1553005
Alumina Neutral, Active, is used for packing flash chromatography columns. It has a particle distribution of 32-63um and a quantity of 500g. The approximate pH is 7.4, Loss on Ignition (LOI) is < 1.7%, and specific surface area is about 200 m2/g.