Learn More
Analytical Sales & Services AluFit Heat Transfer Collar for 10cm Columns
Catalog No. 5000807158
Encompass
AluFit™ Heat Transfer Collar for 10cm Column\n\n• Use for proper heat transfer to analytical size columns when using Hot Sleeve Plus™\n• Provides reduced initial equilibration time
Catalog No. 50-008-07158 Supplier Analytical Sales & Services Supplier No. ALUFIT10
AluFit™ Heat Transfer Collar for 10cm Column\n\n• Use for proper heat transfer to analytical size columns when using Hot Sleeve Plus™\n• Provides reduced initial equilibration time