Learn More
Analytical Sales & Services AluFit Heat Transfer Collar for 25cm Columns
Catalog No. 5000805015
Encompass
AluFit™ Heat Transfer Collar for 25cm Column\n\n• Use for proper heat transfer to analytical size columns when using Hot Sleeve Plus™\n• Provides reduced initial equilibration time
Catalog No. 50-008-05015 Supplier Analytical Sales & Services Supplier No. ALUFIT25
AluFit™ Heat Transfer Collar for 25cm Column\n\n• Use for proper heat transfer to analytical size columns when using Hot Sleeve Plus™\n• Provides reduced initial equilibration time