Learn More
Analytical Sales & Services AluFit Heat Transfer Collar for 30cm Columns
Catalog No. 5000806415
Encompass
AluFit™ Heat Transfer Collar for 30cm Column\n\n• Use for proper heat transfer to analytical size columns when using Hot Sleeve Plus™\n• Provides reduced initial equilibration time
Catalog No. 50-008-06415 Supplier Analytical Sales & Services Supplier No. ALUFIT30
AluFit™ Heat Transfer Collar for 30cm Column\n\n• Use for proper heat transfer to analytical size columns when using Hot Sleeve Plus™\n• Provides reduced initial equilibration time