Learn More
New England Biolabs, Inc. AluI – 5000 units
SDP
Catalog No. 50811998
Encompass
A restriction endonuclease that recognizes the sequence AG_^CT.
Catalog No. 50-811-998 Supplier New England Biolabs, Inc. Supplier No. R0137L
A restriction endonuclease that recognizes the sequence AG_^CT.