Learn More
LabScientific Aluminum Microtube Cooling Rack. Aluminum 200ul PCR Cooling Block (5 x 3 3/8”; 12.7 x 6 cm) 96 well (12 x 8 format)
Catalog No. NC9066571
Encompass
Aluminum Microtube Cooling Rack. Aluminum 200ul PCR Cooling Block (5 x 3 3/8”; 12.7 x 6 cm) 96 well (12 x 8 format)
Catalog No. NC9066571 Supplier LabScientific Supplier No. 2075
Aluminum Microtube Cooling Rack. Aluminum 200ul PCR Cooling Block (5 x 3 3/8”; 12.7 x 6 cm) 96 well (12 x 8 format)