Learn More
Arcticwhite LLC Aluminum Adhesive Sealing Foil Non-Sterile / 100, Low-contamination Adhesive, 100 Sealing Films per Case
SDP
Catalog No. NC9058676
Encompass
Aluminum Adhesive Sealing Foil, Non-Sterile / 100/Pack , Low-contamination Adhesive, 100 Sealing Films per Case. Surface treatment - Low-contamination adhesive. Color - Silver. Temperature range - Wide range, for compound storage and PCR. Supplied by ArcticWhite as 100 sealing films per case.
Catalog No. NC9058676 Supplier Arcticwhite LLC Supplier No. AWSMT3295
Aluminum Adhesive Sealing Foil, Non-Sterile / 100/Pack , Low-contamination Adhesive, 100 Sealing Films per Case. Surface treatment - Low-contamination adhesive. Color - Silver. Temperature range - Wide range, for compound storage and PCR. Supplied by ArcticWhite as 100 sealing films per case.