Learn More
MSC RivetKing Blind Rivet, Open End, Aluminum, Dome Head, 50 Pack
Catalog No. 502866187
Encompass
This open end blind rivet features a dome head and is constructed with an aluminum body and an aluminum mandrel. It has a 3/16 in head diameter and a 3/16 in body diameter, suitable for a grip range of 0.137 to 0.354 in. The product is sold in packages of 50.
  • Open end rivet type
  • Dome head
  • Aluminum body material
  • Aluminum mandrel material
  • Head diameter of 3/16 in
  • Body diameter of 3/16 in
  • Grip range is 0.137–0.354 in
  • Length under head is 0.9300 in
  • Country of origin Taiwan
  • Supplied in 50-pack quantities
Catalog No. 50-286-6187 Supplier MSC Supplier No. 70975826
This open end blind rivet features a dome head and is constructed with an aluminum body and an aluminum mandrel. It has a 3/16 in head diameter and a 3/16 in body diameter, suitable for a grip range of 0.137 to 0.354 in. The product is sold in packages of 50.
  • Open end rivet type
  • Dome head
  • Aluminum body material
  • Aluminum mandrel material
  • Head diameter of 3/16 in
  • Body diameter of 3/16 in
  • Grip range is 0.137–0.354 in
  • Length under head is 0.9300 in
  • Country of origin Taiwan
  • Supplied in 50-pack quantities