Learn More
Instech Aluminum Cap for One Channel Vascular Access Buttons™ for Mice. Package of 5 Different Colors.
SDP
Catalog No. NC2816920
Encompass
Aluminum cap for one channel Vascular Access Buttons™ for mice, 5 colors.
Magnet allows for quick and easy attachment while holding the cap securely in place. Allows for group housing - a recommended animal welfare refinement.
Catalog No. NC2816920 Supplier Instech Supplier No. VABM1C5
Aluminum cap for one channel Vascular Access Buttons™ for mice, 5 colors.
Magnet allows for quick and easy attachment while holding the cap securely in place. Allows for group housing - a recommended animal welfare refinement.