Learn More
Instech Protective Aluminum Caps for Oval Magnetic 2 Channel Mouse VAB™, Package of 5 Different Colors
SDP
Catalog No. NC2816925
Encompass
Aluminum cap for two channel Vascular Access Buttons™ for mice, 5 colors. Magnet allows for quick and easy attachment while holding the cap securely in place. Allows for group housing - a recommended animal welfare refinement.
Catalog No. NC2816925 Supplier Instech Supplier No. VABM2JC5
Aluminum cap for two channel Vascular Access Buttons™ for mice, 5 colors. Magnet allows for quick and easy attachment while holding the cap securely in place. Allows for group housing - a recommended animal welfare refinement.