Learn More
Instech Red aluminum cap for Vascular Access Button™ for mice (VAB62BS/22 and VAB62BS/25). Protects button when animals are group housed. Sterile.
SDP
Catalog No. 50890169
Encompass
The Instech VAB62CAP is a protective red aluminum cap designed for use with non-magnetic mouse Vascular Access Buttons, shielding the port during group housing to prevent damage or contamination. Its durable red anodized aluminum construction provides reliable mechanical protection while maintaining compatibility with both 22ga and 25ga mouse VAB models. Supplied sterile, the cap offers a simple and effective way to safeguard VAB implants between dosing or sampling sessions.
Features
  • Protective cap for non-magnetic mouse VABs
  • Compatible with VAB62BS/22 and VAB62BS/25
  • Anodized red aluminum construction
  • Supports group housing of catheterized mice
  • Sterile, ready to use
Catalog No. 50-890-169 Supplier Instech Supplier No. VAB62CAP
The Instech VAB62CAP is a protective red aluminum cap designed for use with non-magnetic mouse Vascular Access Buttons, shielding the port during group housing to prevent damage or contamination. Its durable red anodized aluminum construction provides reliable mechanical protection while maintaining compatibility with both 22ga and 25ga mouse VAB models. Supplied sterile, the cap offers a simple and effective way to safeguard VAB implants between dosing or sampling sessions.
Features
  • Protective cap for non-magnetic mouse VABs
  • Compatible with VAB62BS/22 and VAB62BS/25
  • Anodized red aluminum construction
  • Supports group housing of catheterized mice
  • Sterile, ready to use