Learn More
Instech Red aluminum cap for Vascular Access Button™ for rats (VAB95BS). Protects button when animals are group housed. Sterile.
SDP
Catalog No. 5081238
Encompass
The Instech VAB95CAP is a sterile red aluminum protective cap designed for use with rat Vascular Access Buttons in the VAB95 series. It securely covers the external port to shield the septum and connector when animals are group housed, helping maintain catheter patency and preventing contamination during periods when the button is not tethered.
Features
  • Sterile protective cap for VAB95‑series rat buttons
  • Red anodized aluminum construction
  • Protects the septum and connector when untethered
  • Supports safe group housing of implanted animals
  • Designed for non‑magnetic and MRI‑compatible VAB95 buttons
Catalog No. 50-812-38 Supplier Instech Supplier No. VAB95CAP
The Instech VAB95CAP is a sterile red aluminum protective cap designed for use with rat Vascular Access Buttons in the VAB95 series. It securely covers the external port to shield the septum and connector when animals are group housed, helping maintain catheter patency and preventing contamination during periods when the button is not tethered.
Features
  • Sterile protective cap for VAB95‑series rat buttons
  • Red anodized aluminum construction
  • Protects the septum and connector when untethered
  • Supports safe group housing of implanted animals
  • Designed for non‑magnetic and MRI‑compatible VAB95 buttons