Learn More
MSC Noga Indicator Positioner, Aluminum, Articulated Arm, Fine Adjustment, 1 ea
Catalog No. 5000510619
Encompass
This articulated arm indicator positioner and holder is designed for precise measurement applications, offering high flexibility for various positioning requirements. Constructed from aluminum with an anti-rust black finish, it provides durability and stability for industrial measuring tasks. The assembly features a central locking mechanism for instant, secure positioning.
  • Features a central locking mechanism with a spring-loaded bearing
  • Equipped with top fine adjustment including an additional hermetic lock to prevent movement during use
  • Universal swivel clamp accommodates 6 mm, 8 mm, 3/8 in. diameters, and dovetail connections
  • Includes a 8 mm x 40 mm centering rod
  • Features a 3-joint articulated arm design for enhanced reach and versatility
  • Total length of 282 mm
Catalog No. 50-005-10619 Supplier MSC Supplier No. 72695380
This articulated arm indicator positioner and holder is designed for precise measurement applications, offering high flexibility for various positioning requirements. Constructed from aluminum with an anti-rust black finish, it provides durability and stability for industrial measuring tasks. The assembly features a central locking mechanism for instant, secure positioning.
  • Features a central locking mechanism with a spring-loaded bearing
  • Equipped with top fine adjustment including an additional hermetic lock to prevent movement during use
  • Universal swivel clamp accommodates 6 mm, 8 mm, 3/8 in. diameters, and dovetail connections
  • Includes a 8 mm x 40 mm centering rod
  • Features a 3-joint articulated arm design for enhanced reach and versatility
  • Total length of 282 mm