Learn More
Perkin Elmer US LLC Autosampler Aluminum Sample Covers - Qty. 400
Catalog No. 509051968
Encompass_Preferred
These Autosampler Aluminum Sample Covers have a length of 10 mm, a depth of 0.55 mm, and a maximum pressure threshold of 1 bar. Compatible with the DSC 4/6/7/Jade DSC/Diamond DSC/ PYRIS 1 DSC/PYRIS 6 DSC. Qty. 400 covers
Catalog No. 50-905-1968 Supplier Perkin Elmer US LLC Supplier No. B0143003
These Autosampler Aluminum Sample Covers have a length of 10 mm, a depth of 0.55 mm, and a maximum pressure threshold of 1 bar. Compatible with the DSC 4/6/7/Jade DSC/Diamond DSC/ PYRIS 1 DSC/PYRIS 6 DSC. Qty. 400 covers