Learn More
METTLER TOLEDO 3P INDUSTRIAL Aluminum crucible standard 40uL with lid
Catalog No. NC1773401
Encompass_Preferred
Aluminum is the standard DSC crucible material up to 640°C. For solids, liquids and powder samples and excellent DSC performance.
Catalog No. NC1773401 Supplier METTLER TOLEDO 3P INDUSTRIAL Supplier No. 26763
Aluminum is the standard DSC crucible material up to 640°C. For solids, liquids and powder samples and excellent DSC performance.