Learn More
LABORATORY SALES AND SERVICE ALUM DISH 57MM PK/100
SDP
Catalog No. NC2596375
Encompass
Disposable smooth wall aluminum weighing dishes are oil-free and noncoated. They are ideal for drying applications, weighting and general purpose sampling.
Catalog No. NC2596375 Supplier LABORATORY SALES AND SERVICE Supplier No. LS1041107PK
Disposable smooth wall aluminum weighing dishes are oil-free and noncoated. They are ideal for drying applications, weighting and general purpose sampling.