Learn More
Electron Microscopy Sciences Alum Hematoxylin Solution 500 ML
SDP
Catalog No. 501901632
Encompass_Preferred
Alum Celestine Blue (Iron Alum – Celestine Blue) solution is a prepared, ready-to-use, high quality staining solution that saves time and effort and produces consistent uniform results. It is a substitute for Hematoxylin/Eosin. See Lendrum & McFarlane, (1940). J. of Pathology & Bacteriology, v. 50, pp 381
Catalog No. 50-190-1632 Supplier Electron Microscopy Sciences Supplier No. 2602305
Alum Celestine Blue (Iron Alum – Celestine Blue) solution is a prepared, ready-to-use, high quality staining solution that saves time and effort and produces consistent uniform results. It is a substitute for Hematoxylin/Eosin. See Lendrum & McFarlane, (1940). J. of Pathology & Bacteriology, v. 50, pp 381