Learn More
Andwin Scientific ALUM.MICRO WEIGH DISH 250/PK
SDP
Catalog No. NC9038456
Encompass_Preferred
The aluminum micro weighing dish is suitable for microbalances, organic micro elementary and thermal analysis, and pharmaceutical use. This product has dimensions of 10W x 4L x 4.5D mm and has a capacity of 0.11 mL. These dishes can be used with most commercially available analyzers and microbalances. It is sold in a pack of 250.
Catalog No. NC9038456 Supplier Andwin Scientific Supplier No. 12577066
The aluminum micro weighing dish is suitable for microbalances, organic micro elementary and thermal analysis, and pharmaceutical use. This product has dimensions of 10W x 4L x 4.5D mm and has a capacity of 0.11 mL. These dishes can be used with most commercially available analyzers and microbalances. It is sold in a pack of 250.