Learn More
Andwin Scientific ALUM MICRO WEIGH DISHES 25X7X7
SDP
Catalog No. NC9452856
Encompass_Preferred
The aluminum micro weighing dish is suitable for microbalances, organic micro elementary and thermal analysis, and pharmaceutical use. This product has dimensions of 25W x 7L x 7D mm and has a capacity of 1.12 mL. These dishes can be used with most commercially available analyzers and microbalances. It is sold in a pack of 50.
Catalog No. NC9452856 Supplier Andwin Scientific Supplier No. 12577068
The aluminum micro weighing dish is suitable for microbalances, organic micro elementary and thermal analysis, and pharmaceutical use. This product has dimensions of 25W x 7L x 7D mm and has a capacity of 1.12 mL. These dishes can be used with most commercially available analyzers and microbalances. It is sold in a pack of 50.