Learn More
Cole-Parmer Essentials Brown Aluminum Oxide, 25 lb
Supplier:  Cole-Parmer 0118463
Encompass_Preferred
Aluminum oxide is ideal for use as a an agent in fluidized beds for heat transfer and maintaining precise temperature control. It is also a fast-cutting abrasive in the range of 80 HRC. The brown aluminum oxide is high-quality fused virgin material produced to conform to all major industrial and governmental standards. The white aluminum oxide is specially treated for the removal of iron in order to prevent contamination of the object being blasted.
Catalog No. NC1591986