Learn More
Perkin Elmer US LLC Sealed Autosampler Aluminum Sample Pans - 30 µL, Qty. 400
Catalog No. 509051971
Encompass_Preferred
These volatile pans and covers are suitable for use with DSC autosamplers, but are also suitable for use with non-robotic systems. Most of these pans and covers can be crimped using the Universal Crimper (B0139005).
Catalog No. 50-905-1971 Supplier Perkin Elmer US LLC Supplier No. B0143016
These volatile pans and covers are suitable for use with DSC autosamplers, but are also suitable for use with non-robotic systems. Most of these pans and covers can be crimped using the Universal Crimper (B0139005).