Learn More
Andwin Scientific ALUM. PAN DISP. 3INWGH PK/50
SDP
Catalog No. NC9303352
Encompass_Preferred
The disposable aluminum weighing pans have several purposes, including but not limited to dispensing, weighing, storing, and drying. The oil-free pans feature a strong aluminum construction and great compatibility with most balances and dryers. This product has a capacity of 40 mL and is sold in a pack of 50.
Catalog No. NC9303352 Supplier Andwin Scientific Supplier No. 12177305
The disposable aluminum weighing pans have several purposes, including but not limited to dispensing, weighing, storing, and drying. The oil-free pans feature a strong aluminum construction and great compatibility with most balances and dryers. This product has a capacity of 40 mL and is sold in a pack of 50.