Learn More
MSC ALUM POLYCARB FACESHIELD
Catalog No. 5000793401
Encompass
This clear face shield window is 9 in. high and 0.08 in. thick, crafted from polycarbonate for durability. It is designed for use with hard hats and meets rigorous safety standards, including ANSI Z87.1 and CSA Z94.3.
  • Designed for use with hard hats
  • Meets CSA Z94.3 standards
  • Meets ANSI Z87.1 standards
  • Made from polycarbonate material
  • Suitable for applications such as welding, sanding, and grinding
Catalog No. 50-007-93401 Supplier MSC Supplier No. 90299868
This clear face shield window is 9 in. high and 0.08 in. thick, crafted from polycarbonate for durability. It is designed for use with hard hats and meets rigorous safety standards, including ANSI Z87.1 and CSA Z94.3.
  • Designed for use with hard hats
  • Meets CSA Z94.3 standards
  • Meets ANSI Z87.1 standards
  • Made from polycarbonate material
  • Suitable for applications such as welding, sanding, and grinding