Learn More
Gilson Company Inc 24oz Aluminum Round Bottom Sample Scoop
Catalog No. NC1875903
Encompass_Preferred
This 24oz (710ml) TSA-186 Sample Scoop with round bottom is constructed of aluminum and is useful for lab or field handling of sample materials. Scoop bowl is 7.0x3.8in (177.8x96.52mm). The overall length is 10.5in (266.7mm). A rigid sure-grip handle prevents slippage.
Catalog No. NC1875903 Supplier Gilson Company Inc Supplier No. TSA186
This 24oz (710ml) TSA-186 Sample Scoop with round bottom is constructed of aluminum and is useful for lab or field handling of sample materials. Scoop bowl is 7.0x3.8in (177.8x96.52mm). The overall length is 10.5in (266.7mm). A rigid sure-grip handle prevents slippage.