Learn More
Cole-Parmer Essentials Aluminum Smooth-Walled Weighing Dishes, 80 mL, 100/Pk
Catalog No. NC2123827 Shop All Cole-Parmer Products
Encompass_Preferred
These smooth-sided, flat bottom, multipurpose "utility" dishes are ideal for storage, dispensing, general purpose sampling, and testing. Dish contour facilitates stacking and dispensing. May be used in a wide variety of applications including weighing, moisture testing, wastewater analysis, chemical, and pharmaceutical. The dishes are oil-free and non-coated.
Catalog No. NC2123827 Supplier Cole-Parmer Supplier No. 0101828
These smooth-sided, flat bottom, multipurpose "utility" dishes are ideal for storage, dispensing, general purpose sampling, and testing. Dish contour facilitates stacking and dispensing. May be used in a wide variety of applications including weighing, moisture testing, wastewater analysis, chemical, and pharmaceutical. The dishes are oil-free and non-coated.