Learn More
MSC Miller Tripod, High-Strength Aluminum, 7 ft., 400 lb. Capacity, 1 ea.
Catalog No. 5000508426
Encompass
This 7 ft. (2 m) high-strength aluminum tripod serves as a portable anchorage connector for confined space entry and rescue operations. Constructed from lightweight aluminum, this system is designed to provide secure support for workers entering, escaping, or working within enclosed spaces.
  • Maximum working load of 400 lbs. (181 kg)
  • Vertical pull capacity of 5,000 lbs. (22 kN)
  • Adjustable legs with integral push pins for uneven surfaces
  • Independent leg locking feature for added security
  • Tripod feet with rubber skid pads and serrated edges to prevent slippage
  • Includes safety chain and upper anchorage points
  • Meets OSHA 1926.502 and ANSI Z359.1 standards
Catalog No. 50-005-08426 Supplier MSC Supplier No. 52267218
This 7 ft. (2 m) high-strength aluminum tripod serves as a portable anchorage connector for confined space entry and rescue operations. Constructed from lightweight aluminum, this system is designed to provide secure support for workers entering, escaping, or working within enclosed spaces.
  • Maximum working load of 400 lbs. (181 kg)
  • Vertical pull capacity of 5,000 lbs. (22 kN)
  • Adjustable legs with integral push pins for uneven surfaces
  • Independent leg locking feature for added security
  • Tripod feet with rubber skid pads and serrated edges to prevent slippage
  • Includes safety chain and upper anchorage points
  • Meets OSHA 1926.502 and ANSI Z359.1 standards