Learn More
Cole-Parmer Cole-Parmer Micro Aluminum Weighing Dishes, 25mm L x 7mm W x 7mm H, 1.12 mL, 50/pk
Catalog No. NC0880763 Shop All Cole-Parmer Products
Encompass_Preferred
Ideal for microbalances, organic micro elementary and thermal analysis, and pharmaceutical use. Our micro weigh boats are compatible with most commercially available analyzers and micro balances. Packaged in easy-to-use plastic containers for storage and dispensing. The dime is not included.
Catalog No. NC0880763 Supplier Cole-Parmer Supplier No. 0101901
Ideal for microbalances, organic micro elementary and thermal analysis, and pharmaceutical use. Our micro weigh boats are compatible with most commercially available analyzers and micro balances. Packaged in easy-to-use plastic containers for storage and dispensing. The dime is not included.