Learn More
Cole-Parmer Cole-Parmer 43 mm Aluminum Crimpled-Walled Weighing Dishes with Tab, 20 mL, 100/Pk
Catalog No. NC1784817 Shop All Cole-Parmer Products
Encompass_Preferred
Crimped sides for rigidity, sturdy gauge for heavy duty use. Large tab for sample numbers, flat bottom to improve sample handling, sides are angled for easy seperation. No oil residue, no vinyl coating to create contamination problems. Smooth bottom and may be used in a wide variety of applications including weighing, moisture testing, wastewater analysis, chemical, and pharmaceutical.
Catalog No. NC1784817 Supplier Cole-Parmer Supplier No. 0101851
Crimped sides for rigidity, sturdy gauge for heavy duty use. Large tab for sample numbers, flat bottom to improve sample handling, sides are angled for easy seperation. No oil residue, no vinyl coating to create contamination problems. Smooth bottom and may be used in a wide variety of applications including weighing, moisture testing, wastewater analysis, chemical, and pharmaceutical.