Learn More
Andwin Scientific ALUM WEIGH DISH 47MM 144/PK
SDP
Catalog No. NC9040422
Encompass_Preferred
The aluminum crinkle dishes are supreme products for weighing, dispensing, and storing purposes. The dishes can also be utilized as vaporating dishes, parts containers, or dust covers.This product is specially designed with tabs that allow easy and convenient handling and crimped sides for sturdiness. This product has a capacity of 60 mL or 2 oz and is sold in a pack of 144.
Catalog No. NC9040422 Supplier Andwin Scientific Supplier No. 25433008PK
The aluminum crinkle dishes are supreme products for weighing, dispensing, and storing purposes. The dishes can also be utilized as vaporating dishes, parts containers, or dust covers.This product is specially designed with tabs that allow easy and convenient handling and crimped sides for sturdiness. This product has a capacity of 60 mL or 2 oz and is sold in a pack of 144.