Learn More
Andwin Scientific ALUM WEIGH PAN 5IN DIA PK/100
SDP
Catalog No. NC9762670
Encompass_Preferred
The disposable aluminum weighing pans have several purposes, including but not limited to dispensing, weighing, storing, and drying. The oil-free pans feature a strong aluminum construction and great compatibility with most balances and dryers. This product has a capacity of 50 mL and is sold in a pack of 100.
Catalog No. NC9762670 Supplier Andwin Scientific Supplier No. 12175001
The disposable aluminum weighing pans have several purposes, including but not limited to dispensing, weighing, storing, and drying. The oil-free pans feature a strong aluminum construction and great compatibility with most balances and dryers. This product has a capacity of 50 mL and is sold in a pack of 100.