Learn More
Andwin Scientific ALUMA SEAL TAPE 96 100/PK
SDP
Catalog No. NC9523830
Encompass
The AlumaSeal sealing film is ideal for use with 96-well plates. The film can fit inside the rim of raised-rim plates and can withstand a temperature range of -80 C to +120 C. This product is free of DNase, RNase, and nucleic acids.
Catalog No. NC9523830 Supplier Andwin Scientific Supplier No. F96100
The AlumaSeal sealing film is ideal for use with 96-well plates. The film can fit inside the rim of raised-rim plates and can withstand a temperature range of -80 C to +120 C. This product is free of DNase, RNase, and nucleic acids.